Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Actn4
Entrez Gene ID
63836
Gene Name
actinin alpha 4
Gene Aliases
-
Chromosome Location
Chr.1: 87078012 - 87147347 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031675.2 NP_113863.2 20 - 21 2678 92
XM_006228687.1 XP_006228749.1 20 - 21 2717 92
XM_006228688.1 XP_006228750.1 20 - 21 2702 92
XM_006228689.1 XP_006228751.1 20 - 21 2702 92
XM_006228690.2 XP_006228752.1 20 - 21 2996 92
XM_008759185.1 XP_008757407.1 21 - 22 2783 92
GenBank mRNA AY724534.1 - 20 - 21 2647 92
BC061788.1 - 20 - 21 2669 92
U19893.1 - 20 - 21 2679 92

View Full Product Details