Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gltscr2
Entrez Gene ID
292624
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
-
Chromosome Location
Chr.1: 77836522 - 77843991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207591.1 NP_997474.1 11 - 12 1388 89
GenBank mRNA AY535002.1 - 11 - 12 1388 89
BC086359.1 - 11 - 12 1392 89
FQ212888.1 - 11 - 12 1423 89
FQ213201.1 - 11 - 12 1397 89
FQ215499.1 - 9 - 10 1089 89
FQ219058.1 - 11 - 12 1395 89
FQ222789.1 - 11 - 12 1398 89

View Full Product Details