Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gltscr2
Entrez Gene ID
292624
Gene Name
glioma tumor suppressor candidate region gene 2
Gene Aliases
-
Chromosome Location
Chr.1: 77836522 - 77843991 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC086359.1 - 12 - 13 1448 60
FQ212888.1 - 12 - 13 1479 60
FQ213201.1 - 12 - 13 1453 60
FQ215499.1 - 10 - 11 1145 60
FQ218937.1 - 12 - 13 1451 60
FQ219058.1 - 12 - 13 1451 60

View Full Product Details