Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rilpl1
Entrez Gene ID
304469
Gene Name
Rab interacting lysosomal protein-like 1
Gene Aliases
GOSPEL, RGD1307973
Chromosome Location
Chr.12: 37490320 - 37528395 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249294.1 XP_006249356.1 6 - 7 1542 140

View Full Product Details