The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009679.1 | NP_001009679.1 | 5 - 6 | 803 | 62 |
GenBank mRNA | BC088310.1 | - | 5 - 6 | 803 | 62 |
FQ213675.1 | - | 5 - 6 | 794 | 62 |