Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zp3
Entrez Gene ID
114639
Gene Name
zona pellucida glycoprotein 3 (sperm receptor)
Gene Aliases
Zp-3
Chromosome Location
Chr.12: 23735225 - 23742190 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC127488.1 - 6 - 7 961 123

View Full Product Details