Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sbds
Entrez Gene ID
288615
Gene Name
Shwachman-Bodian-Diamond syndrome
Gene Aliases
-
Chromosome Location
Chr.12: 29921443 - 29930602 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008289.1 NP_001008290.1 1 - 2 153 83
GenBank mRNA BC086335.1 - 1 - 2 153 83

View Full Product Details