Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wsb2
Entrez Gene ID
288692
Gene Name
WD repeat and SOCS box-containing 2
Gene Aliases
-
Chromosome Location
Chr.12: 44941655 - 44962679 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007616.1 NP_001007617.1 8 - 9 1116 75
XM_008769220.1 XP_008767442.1 7 - 8 1053 75
GenBank mRNA BC081901.1 - 8 - 9 1116 75

View Full Product Details