Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oasl
Entrez Gene ID
304545
Gene Name
2'-5'-oligoadenylate synthetase-like
Gene Aliases
Oasl1
Chromosome Location
Chr.12: 47444138 - 47456160 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009681.1 NP_001009681.1 1 - 2 206 64
GenBank mRNA AY227756.1 - 1 - 2 206 64

View Full Product Details