Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhof
Entrez Gene ID
690130
Gene Name
ras homolog family member F (in filopodia)
Gene Aliases
-
Chromosome Location
Chr.12: 38879762 - 38894095 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001073389.5 XP_001073389.2 1 - 2 833 68

View Full Product Details