Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rnft2
Entrez Gene ID
304521
Gene Name
ring finger protein, transmembrane 2
Gene Aliases
RGD1560195, Tmem118
Chromosome Location
Chr.12: 43940929 - 44008692 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107144.1 NP_001100614.1 8 - 9 1330 116
XM_008769232.1 XP_008767454.1 8 - 9 1291 116

View Full Product Details