Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wscd2
Entrez Gene ID
360824
Gene Name
WSC domain containing 2
Gene Aliases
Wscd2-ps1
Chromosome Location
Chr.12: 48817320 - 48857724 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271178.1 NP_001258107.1 7 - 8 1481 106

View Full Product Details