Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Caln1
Entrez Gene ID
363909
Gene Name
calneuron 1
Gene Aliases
-
Chromosome Location
Chr.12: 29307117 - 29743705 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249243.2 XP_006249305.1 1 - 2 88 67

View Full Product Details