Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem255b
Entrez Gene ID
290877
Gene Name
transmembrane protein 255B
Gene Aliases
Fam70b, RGD1566042
Chromosome Location
Chr.16: 81182478 - 81209429 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106094.2 NP_001099564.1 2 - 3 276 88
XM_008771387.1 XP_008769609.1 8 - 9 1024 88
XM_008771388.1 XP_008769610.1 7 - 8 963 88
XM_008771389.1 XP_008769611.1 7 - 8 935 88
GenBank mRNA BC169123.1 - 2 - 3 276 88

View Full Product Details