Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubxn8
Entrez Gene ID
290802
Gene Name
UBX domain protein 8
Gene Aliases
RGD1306023, Ubxd6
Chromosome Location
Chr.16: 62251243 - 62272838 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106086.1 NP_001099556.1 3 - 4 1694 74

View Full Product Details