Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam90a1
Entrez Gene ID
100362477
Gene Name
family with sequence similarity 90, member A1
Gene Aliases
Fam90a1-ps2, Fam90a13p
Chromosome Location
Chr.16: 75001254 - 75016168 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728417.3 XP_002728463.2 2 - 3 445 99

View Full Product Details