Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nek5
Entrez Gene ID
685440
Gene Name
NIMA-related kinase 5
Gene Aliases
-
Chromosome Location
Chr.16: 74809513 - 74851895 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771370.1 XP_008769592.1 3 - 4 297 94

View Full Product Details