Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc20a2
Entrez Gene ID
29502
Gene Name
solute carrier family 20 (phosphate transporter), member 2
Gene Aliases
Ab1-188, Glvr2, Pit-2, RAM-1, Ram1
Chromosome Location
Chr.16: 74318287 - 74408030 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771351.1 XP_008769573.1 1 - 2 1154 121

View Full Product Details