Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erlin2
Entrez Gene ID
290823
Gene Name
ER lipid raft associated 2
Gene Aliases
RGD1309010, Spfh2
Chromosome Location
Chr.16: 69179005 - 69195458 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106088.2 NP_001099558.1 5 - 6 343 138
XM_008771323.1 XP_008769545.1 5 - 6 596 138
XM_008771324.1 XP_008769546.1 5 - 6 538 138
GenBank mRNA BC168668.1 - 5 - 6 343 138

View Full Product Details