Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mfhas1
Entrez Gene ID
306508
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
-
Chromosome Location
Chr.16: 60120457 - 60208545 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253224.2 XP_006253286.1 3 - 3 5749 64
XM_006253225.2 XP_006253287.1 4 - 4 5859 64
GenBank mRNA FQ232773.1 - 1 - 1 728 64

View Full Product Details