The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134744.1 | NP_001128216.1 | 7 - 8 | 869 | 106 |
XM_006253381.2 | XP_006253443.1 | 7 - 8 | 1141 | 106 | |
GenBank mRNA | BC168222.1 | - | 7 - 8 | 869 | 106 |