Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ascl5
Entrez Gene ID
363991
Gene Name
achaete-scute family bHLH transcription factor 5
Gene Aliases
RGD1561854
Chromosome Location
Chr.13: 52887649 - 52888215 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769577.1 XP_008767799.1 1 - 1 145 67

View Full Product Details