Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Igfn1
Entrez Gene ID
304823
Gene Name
immunoglobulin-like and fibronectin type III domain containing 1
Gene Aliases
Igfn1-ps1, RGD1565384
Chromosome Location
Chr.13: 52791188 - 52821001 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008769584.1 XP_008767806.1 14 - 15 6343 82

View Full Product Details