Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trove2
Entrez Gene ID
304833
Gene Name
TROVE domain family, member 2
Gene Aliases
Ssa2
Chromosome Location
Chr.13: 60546892 - 60567882 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107183.1 NP_001100653.1 8 - 9 1604 71

View Full Product Details