The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107179.1 | NP_001100649.1 | 1 - 2 | 464 | 128 |
XM_008769562.1 | XP_008767784.1 | 1 - 2 | 892 | 128 |