The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134361.1 | NP_599188.1 | 2 - 3 | 177 | 81 |
GenBank mRNA | U23377.1 | - | 2 - 3 | 177 | 81 |