Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl13
Entrez Gene ID
289159
Gene Name
methyltransferase like 13
Gene Aliases
RGD1311526
Chromosome Location
Chr.13: 80427595 - 80441352 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134501.1 NP_001127973.1 3 - 4 1381 97
XM_006250152.2 XP_006250214.1 4 - 5 1327 97
XM_008769622.1 XP_008767844.1 3 - 4 1429 97

View Full Product Details