Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx9
Entrez Gene ID
289048
Gene Name
LIM homeobox 9
Gene Aliases
-
Chromosome Location
Chr.13: 55854543 - 55879305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006249918.2 XP_006249980.1 4 - 5 1518 79
XM_006249919.2 XP_006249981.1 7 - 8 1593 79
XM_008769545.1 XP_008767767.1 6 - 7 1119 79
GenBank mRNA BC128722.1 - 7 - 8 1107 79

View Full Product Details