The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106498.1 | NP_001099968.1 | 10 - 11 | 2517 | 67 |
XM_008762091.1 | XP_008760313.1 | 9 - 10 | 3817 | 67 |