Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rag1
Entrez Gene ID
84600
Gene Name
recombination activating gene 1
Gene Aliases
-
Chromosome Location
Chr.3: 91206394 - 91217491 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053468.1 NP_445920.1 1 - 2 85 110

View Full Product Details