Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve19
Entrez Gene ID
499871
Gene Name
zinc finger, FYVE domain containing 19
Gene Aliases
-
Chromosome Location
Chr.3: 111013856 - 111021846 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034948.1 NP_001030120.1 6 - 7 682 69
XM_008762106.1 XP_008760328.1 6 - 7 682 69
GenBank mRNA BC099795.1 - 6 - 7 682 69

View Full Product Details