Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc73
Entrez Gene ID
499848
Gene Name
coiled-coil domain containing 73
Gene Aliases
RGD1563448
Chromosome Location
Chr.3: 94848771 - 94989983 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234667.2 XP_006234729.1 12 - 13 1223 66

View Full Product Details