The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134834.1 | NP_001128306.1 | 6 - 7 | 952 | 92 |
XM_006234960.2 | XP_006235022.1 | 6 - 7 | 970 | 92 | |
GenBank mRNA | BC166933.1 | - | 6 - 7 | 952 | 92 |