Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Emc4
Entrez Gene ID
296049
Gene Name
ER membrane protein complex subunit 4
Gene Aliases
RGD1310905, Tmem85
Chromosome Location
Chr.3: 103852139 - 103858423 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106495.1 NP_001099965.1 2 - 3 324 55
GenBank mRNA FQ215850.1 - 2 - 3 264 55
FQ234005.1 - 2 - 3 264 55

View Full Product Details