The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005556.1 | NP_001005556.1 | 6 - 7 | 1387 | 68 |
XM_006235034.2 | XP_006235096.1 | 6 - 7 | 1255 | 68 | |
XM_006235035.2 | XP_006235097.1 | 6 - 7 | 1969 | 68 | |
XM_006235036.2 | XP_006235098.1 | 7 - 8 | 1288 | 68 | |
XM_006235037.2 | XP_006235099.1 | 5 - 6 | 1291 | 68 | |
XM_008762179.1 | XP_008760401.1 | 6 - 7 | 2462 | 68 | |
XM_008762180.1 | XP_008760402.1 | 6 - 7 | 1275 | 68 | |
GenBank mRNA | BC081869.1 | - | 6 - 7 | 1387 | 68 |