Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alkbh3
Entrez Gene ID
362169
Gene Name
alkB, alkylation repair homolog 3 (E. coli)
Gene Aliases
-
Chromosome Location
Chr.3: 82874242 - 82906656 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014180.1 NP_001014202.1 1 - 2 64 74
GenBank mRNA BC083756.1 - 1 - 2 64 74

View Full Product Details