Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
295674
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
-
Chromosome Location
Chr.3: 63536166 - 63557378 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013077.1 NP_001013095.1 6 - 7 916 96
GenBank mRNA BC083759.1 - 6 - 7 916 96

View Full Product Details