Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnks1bp1
Entrez Gene ID
295707
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.3: 72458539 - 72483206 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006234484.2 XP_006234546.1 6 - 7 4406 107
XM_006234485.2 XP_006234547.1 6 - 7 4886 107
XM_006234486.2 XP_006234548.1 6 - 7 4349 107

View Full Product Details