The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106497.1 | NP_001099967.1 | 4 - 5 | 316 | 66 |
GenBank mRNA | BC166931.1 | - | 4 - 5 | 272 | 66 |
FQ210704.1 | - | 3 - 4 | 216 | 66 | |
FQ210780.1 | - | 3 - 4 | 163 | 66 |