Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcp11l1
Entrez Gene ID
499846
Gene Name
t-complex 11, testis-specific-like 1
Gene Aliases
RGD1562313
Chromosome Location
Chr.3: 94622403 - 94657467 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109202.1 NP_001102672.1 2 - 3 376 72

View Full Product Details