Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc1a2
Entrez Gene ID
29482
Gene Name
solute carrier family 1 (glial high affinity glutamate transporter), member 2
Gene Aliases
Eaat2, Glt, Glt-1
Chromosome Location
Chr.3: 92518679 - 92665731 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035233.1 NP_001030310.1 - 1386 92
NM_017215.2 NP_058911.2 - 1386 92
GenBank mRNA AF297648.1 - - 1298 92
AF451299.1 - - 758 92
AF465909.1 - - 1279 92
AY044832.1 - - 1275 92
AY069978.1 - - 577 92
JN132400.1 - - 1461 92
U15098.1 - - 1335 92
X67857.1 - - 1373 92

View Full Product Details