Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fsip2
Entrez Gene ID
680315
Gene Name
fibrous sheath-interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 70285181 - 70356568 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761999.1 XP_008760221.1 1 - 2 205 61

View Full Product Details