Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spata5l1
Entrez Gene ID
691729
Gene Name
spermatogenesis associated 5-like 1
Gene Aliases
-
Chromosome Location
Chr.3: 114747410 - 114760409 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109647.1 NP_001103117.1 8 - 8 2407 108

View Full Product Details