Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crls1
Entrez Gene ID
366196
Gene Name
cardiolipin synthase 1
Gene Aliases
RGD1311037
Chromosome Location
Chr.3: 125503638 - 125522446 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014258.1 NP_001014280.1 6 - 7 951 82
XM_006235078.2 XP_006235140.1 6 - 7 1145 82
XM_006235079.2 XP_006235141.1 6 - 7 1134 82
XM_006235080.2 XP_006235142.1 6 - 7 634 82
GenBank mRNA BC085849.1 - 6 - 7 951 82

View Full Product Details