Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mmp20
Entrez Gene ID
300341
Gene Name
matrix metallopeptidase 20
Gene Aliases
-
Chromosome Location
Chr.8: 5823147 - 5875351 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106800.1 NP_001100270.1 4 - 5 675 71
XM_006242487.2 XP_006242549.1 5 - 6 944 71

View Full Product Details