Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Icam5
Entrez Gene ID
313785
Gene Name
intercellular adhesion molecule 5, telencephalin
Gene Aliases
-
Chromosome Location
Chr.8: 22050222 - 22057209 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172079.1 NP_001165550.1 4 - 5 1189 72

View Full Product Details