Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fdx1l
Entrez Gene ID
313786
Gene Name
ferredoxin 1-like
Gene Aliases
RGD1561264
Chromosome Location
Chr.8: 22086534 - 22091450 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108002.1 NP_001101472.1 1 - 2 133 104

View Full Product Details