Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc67
Entrez Gene ID
315438
Gene Name
coiled-coil domain containing 67
Gene Aliases
RGD1306632
Chromosome Location
Chr.8: 14094742 - 14157235 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014087.1 NP_001014109.2 13 - 14 1756 68
XM_008765933.1 XP_008764155.1 13 - 14 2068 68
XM_008765934.1 XP_008764156.1 13 - 14 2001 68
XM_008765935.1 XP_008764157.1 12 - 13 2110 68
XM_008765936.1 XP_008764158.1 13 - 14 2007 68
GenBank mRNA BC085333.1 - 13 - 14 1756 68

View Full Product Details