Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcun1d5
Entrez Gene ID
315405
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 5
Gene Aliases
RGD1307008
Chromosome Location
Chr.8: 5437071 - 5458152 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009696.1 NP_001009696.1 6 - 7 797 115
GenBank mRNA BC087627.1 - 6 - 7 797 115

View Full Product Details