The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053825.2 | NP_446277.1 | 8 - 9 | 964 | 119 |
XM_006254231.2 | XP_006254293.1 | 8 - 9 | 930 | 119 | |
GenBank mRNA | BC059158.1 | - | 8 - 9 | 968 | 119 |
FQ212651.1 | - | 8 - 9 | 962 | 119 | |
FQ213852.1 | - | 8 - 9 | 964 | 119 | |
FQ213883.1 | - | 8 - 9 | 959 | 119 | |
FQ218995.1 | - | 8 - 9 | 964 | 119 | |
FQ219484.1 | - | 8 - 9 | 962 | 119 | |
FQ229987.1 | - | 8 - 9 | 985 | 119 | |
L19927.1 | - | 6 - 7 | 842 | 119 |